Anti-MDK Rabbit Polyclonal Antibody
Supplier: Biorbyt
Certificates
About this item
Type: Primary
Antigen: MDK (midkine (neurite growth-promoting factor 2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Specifications
- Catalog No:
- BIRBORB129681-100
- BIRBORB129681-200
- Antigen symbol:
- MDK
- MDK
- Antigen name:
- midkine (neurite growth-promoting factor 2)
- midkine (neurite growth-promoting factor 2)
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Immunogen:
- Recombinant Protein
- Recombinant Protein
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together