Anti-IL6 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
BIRBORB124689-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Interleukin 6
- Antigen symbol:IL6
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rabbit
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:24 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: IL6 (interleukin 6 (interferon, beta 2))
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit