Anti-IL6 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # BIRBORB124685-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 6
- Antigen symbol:IL6
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:23 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Epitope mapping near the N-terminus of IL-6 of human
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: IL6 (interleukin 6 (interferon, beta 2))
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human