Anti-FPR1 Rabbit Polyclonal Antibody (Cy7®)
BIRBORB123648-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Formyl Peptide Receptor 1
- Antigen symbol:FPR1
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:39 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: FPR1 (formyl peptide receptor 1)
Clonality: polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat