Anti-DKK3 Rabbit Polyclonal Antibody (Cy5.5®)
BIRBORB123114-100
: Biorbyt
- Antibody type:Primary
- Antigen name:dickkopf WNT signaling pathway inhibitor 3
- Antigen symbol:DKK3
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:38 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic peptide derived from human DKK3.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: DKK3 (dickkopf WNT signaling pathway inhibitor 3)
Clonality: polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat