Anti-DKK1 Rabbit Polyclonal Antibody (Cy5.5®)
Artikelnr. BIRBORB123113-100
Lieferant: Biorbyt
Spezifikationen
- Antibody type:Primary
- Antigen name:dickkopf WNT signaling pathway inhibitor 1
- Antigen symbol:DKK1
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:29 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Spezifikationen
Über diesen Artikel
Type: Primary
Antigen: DKK1 (dickkopf WNT signaling pathway inhibitor 1)
Clonality: polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat