Anti-CITED1 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BIRBORB122582-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 1
- Antigen symbol:CITED1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:20 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic peptide derived from human CITED1/ABCC1.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CITED1 (Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 1)
Clonality: polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat