Anti-hCK2, beta subunit Mouse Monoclonal Antibody
BIRBORB122546-50
: Biorbyt
- Antibody type:Primary
- Antigen symbol:hCK2, beta subunit
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: hCK2, beta subunit
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: