Anti-His tag Rabbit Polyclonal Antibody (Cy3®)
BIRBORB114421-100
: Biorbyt
- Antibody type:Primary
- Antigen name:His-tag
- Antigen symbol:His
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Sequence:HHHHHH
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:C and N terminus
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: His tag (His tag)
Clonality: polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: