Anti-NFE2L2 Rabbit Polyclonal Antibody (Biotin)
BIRBORB114290-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Nuclear factor, erythroid 2-like 2
- Antigen symbol:NFE2L2
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Modification:phospho-Ser40
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:68 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human Nrf2 around the phosphorylation site of(Ser40).
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: NFE2L2 (nuclear factor, erythroid 2-like 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: