Anti-M13 bacteriophage Mouse Monoclonal Antibody
Catalog # BIRBORB109590-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:M13 bacteriophage
- Antigen symbol:M13
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Hybridoma produced by the fusion of splenocytes from mice immunized with isolated M13 phage coat proteins and mouse myeloma cells.
- Purification:purified by Protein A/G Chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: M13 bacteriophage
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: