Anti-Phosphatidic Acid Phosphatase Domain Containing 2 Rabbit Polyclonal Antibody
Catalog # BIRBORB108934-10
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Phosphatidic Acid Phosphatase Domain Containing 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide derived from human phosphatidic acid phosphatase type 2 domain containing 2 (PPAPDC2)
- Size:10 Tests
- Pk:10
Specifications
About this item
Type: Primary
Antigen: Phosphatidic Acid Phosphatase Domain Containing 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human