Anti-L-type calcium channel alpha2/delta subunit Rabbit Polyclonal Antibody
Catalog # BIRBORB108855-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:L-type calcium channel alpha2/delta subunit
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide derived from the rat alpha2/delta calcium channel conjugated to KLH
- Purification:purified by ammonium sulfate precipitation
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: L-type calcium channel alpha2/delta subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: