Anti-ch-Alk2 Sheep Polyclonal Antibody
BIRBORB108832-250
: Biorbyt
- Antibody type:Primary
- Antigen symbol:ch-Alk2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Reactivity:Chicken
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Recombinant protein derived from the extracellular domain of the chicken ALK2 protein. Protein shares significant homology with the mouse, rat and human sequences.
- Purification:purified by ammonium sulfate precipitation
- Size:250 µg
- Pk:250 µG
Type: Primary
Antigen: ch-Alk2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Chicken