Anti-DAP Kinase 3 Rabbit polyclonal antibody
Catalog # BIRBORB108729-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:DAP Kinase 3
- Antigen symbol:DAPK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Antibody raised against a synthetic protein corresponding to amino acids 279 to 298 of the human ZIP kinase protein.
- Purification:purified by antigen immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: DAP Kinase 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat