Anti-Protein Phosphatase 2 A/B gamma Rabbit Polyclonal Antibody
Catalog # BIRBORB108631-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Protein Phosphatase 2 A/B gamma
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Antibody raised against synthetic peptide corresponding to amino acids 53 to 66 of the mammalian protein phosphatase 2 A/B conjugated to KLH.
- Purification:purified by ammonium sulfate precipitation
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Protein Phosphatase 2 A/B gamma
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: