Anti-TNFRSF1A Rabbit Polyclonal Antibody (Atto Dyes and Enzyme-Protein Labels)
Artikelnr. BIRBORB107276-100
Lieferant: Biorbyt
Spezifikationen
- Antibody type:Primary
- Antigen name:tumor necrosis factor receptor superfamily, member 1A
- Antigen symbol:TNFRSF1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:Phosphate-buffered solution 0,09% sodium azide and 0,2% (w/v) BSA
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Peptide corresponding to AA 20-43 of the mouse TNF-R1 sequence, identical to rat and human over those residues
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Spezifikationen
Über diesen Artikel
Type: Primary
Antigen: TNFRSF1A (tumor necrosis factor receptor superfamily, member 1A)
Clonality: polyclonal
Clone:
Conjugation: Atto Dyes and Enzyme-Protein Labels
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat