Anti-IL3 Rabbit Polyclonal Antibody
Catalog # BIRBORB48041-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 3
- Antigen symbol:IL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized
- Storage buffer:0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg NaN3.
- Storage temperature:Cool
- Concentration:500 ug/ml
- Shipping temperature:Ice
- Immunogen:E. coli-derived human IL3 recombinant protein (Position: A20-F152).
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: IL3 (interleukin 3 (colony-stimulating factor, multiple))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human