Anti-TNFRSF10D Mouse Monoclonal Antibody (PE (Phycoerythrin))
BIRBORB44525-0.1
: Biorbyt
- Antibody type:Primary
- Antigen name:tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain
- Antigen symbol:TNFRSF10D
- Clonality:Monoclonal
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: TNFRSF10D (tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human