Anti-TNFRSF10C Mouse Monoclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB44521-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:tumor necrosis factor receptor superfamily, member 10c, decoy without an intracellular domain
- Antigen symbol:TNFRSF10C
- Clonality:Monoclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: TNFRSF10C (tumor necrosis factor receptor superfamily, member 10c, decoy without an intracellular domain)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human