Anti-CD27 Mouse Monoclonal Antibody
Catalog # BIRBORB43949-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:CD27 molecule
- Antigen symbol:CD27
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: CD27 (CD27 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human