Anti-Lambda (L chain) Mouse Monoclonal Antibody
Catalog # BIRBORB43602-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:lambda light chain
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 test
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: Lambda (L chain)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human