Anti-TEAD4 Rabbit Polyclonal Antibody
BIRBORB41486-100
: Biorbyt
- Antibody type:Primary
- Antigen name:TEA domain family member 4
- Antigen symbol:TEAD4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Amino acid number:1 to 230
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 230 of TEAD4.
- Purification:purified by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
- Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:500
Type: Primary
Antigen: TEAD4 (TEA domain family member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human