Anti-cytochrome c oxidase subunit VIa polypeptide 2 precursor Rabbit Polyclonal Antibody
Catalog # BIRBORB41427-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:cytochrome c oxidase subunit VIa polypeptide 2 precursor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Amino acid number:34 to 97
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:0,9 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic peptide contain a sequence corresponding to a region within amino acids 34 and 97 of Human COX6A2.
- Purification:purified by antigen affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Western Blot: 1:500-1:3000
Type: Primary
Antigen: cytochrome c oxidase subunit VIa polypeptide 2 precursor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human