Anti-RIC8B Rabbit Polyclonal Antibody
Catalog # BIRBORB41360-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:RIC8 guanine nucleotide exchange factor B
- Antigen symbol:RIC8B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Amino acid number:478 to 537
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:0,85 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic peptide contain a sequence corresponding to a region within amino acids 478 and 537 of Human RIC8A.
- Purification:purified by antigen affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Western Blot: 1:500-1:3000
Type: Primary
Antigen: RIC8B (RIC8 guanine nucleotide exchange factor B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human