Anti-steroidogenic acute regulatory protein isoform 1 Rabbit Polyclonal Antibody
Catalog # BIRBORB41182-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:steroidogenic acute regulatory Protein isoform 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Amino acid number:15 to 238
- Storage buffer:0,1M Tris, 0,1M Glycine, 20% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 15 and 238 of Human STAR.
- Purification:purified by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Western Blot: 1:500-1:3000
Type: Primary
Antigen: steroidogenic acute regulatory protein isoform 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human