Order Entry
Austria
ContactUsLinkComponent
 
Anti-Pyrralin (Advanced Glycation End Product) Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
  BIRBORB27507-20
 :  Biorbyt
Anti-Pyrralin (Advanced Glycation End Product) Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
  BIRBORB27507-20
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    Pyrralin
  • Clonality:
    Monoclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Format:
    concentrated
  • Form:
    liquid
  • Antigen synonyms:
    Advanced Glycation End Product
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Pyrraline-HSA
  • Size:
    20 µg
  • Pk:
    20 µG

 

 

ELISA: 0.1-0.5 ?g/ml; Immunochemistry: 2 ?g/ml

Type: Primary
Antigen: Pyrralin (Advanced Glycation End Product)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: