Order Entry
Austria
ContactUsLinkComponent
Anti-Pentosidine (Advanced Glycation End Product) Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Anti-Pentosidine (Advanced Glycation End Product) Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB27504-50
Supplier:  Biorbyt
Anti-Pentosidine (Advanced Glycation End Product) Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB27504-50
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Pentosidine (Advanced Glycation End Product)
  • Clonality:
    Monoclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Format:
    concentrated
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Pentosidine-HSA
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

ELISA: 0.1-0.5 ?g/ml; Immunochemistry: 2 ?g/ml

Type: Primary
Antigen: Pentosidine (Advanced Glycation End Product)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: