Anti-EGF Receptor (non-ligand binding site) Mouse monoclonal antibody
Catalog # BIRBORB26480-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:EGF Receptor (non-ligand binding site)
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:1 mg/ml (prior to lyophilization)
- Shipping temperature:Ice
- Immunogen:Human EGF Receptor
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: EGF Receptor (non-ligand binding site)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: