Order Entry
Austria
ContactUsLinkComponent
 
Anti-CD41 gpIIb IIIa Mouse Monoclonal Antibody
  BIRBORB26270-0.2
 :  Biorbyt
Anti-CD41 gpIIb IIIa Mouse Monoclonal Antibody
  BIRBORB26270-0.2
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen symbol:
    CD41 gpIIb IIIa
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    lyophilized
  • Storage buffer:
    PBS with 1 mg/mL BSA.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Human Platelets
  • Size:
    200 μg
  • Pk:
    0,2 mg

 

 

Flow Cytometry: 2 ?g/5x106; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: CD41 gpIIb IIIa
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human