Anti-HLA, Class II-DR+DP Mouse Monoclonal Antibody
BIRBORB26094-100
: Biorbyt
- Antibody type:Primary
- Antigen name:HLA, Class II-DR+DP
- Antigen symbol:HLA DR DP
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:Lot specific
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: HLA, Class II-DR+DP
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: