Anti-CCNE1 Rabbit Polyclonal Antibody
100 µl
Catalog # BIRBORB25975-100
Marketsource
Supplier: Biorbyt
Jump to:
Product Details & Documents
DB: 1:200-1:500; ELISA: 1:2000
Type: Primary
Antigen: CCNE1 (cyclin E1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Documents
orb25975
Datasheet
Specifications
- Size:100 µl
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Storage temperature:Cool
- Host:Rabbit
- Concentration:Total Protein: 85 mg/ml (Refractometry)
- Shipping temperature:Ice
- Antigen name:cyclin E1
- Cat. No.:BIRBORB25975-100
- Immunogen:Full length synthetic peptide corresponding to the human gene sequence.
- Conjugation:Unconjugated
- Antigen symbol:CCNE1
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



