Anti-Kappa Light Chain Goat Polyclonal Antibody
Catalog # BIRBORB25809-10
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Kappa Light Chain
- Antigen symbol:IGKC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Storage buffer:Tris-saline, pH 7,4
- Storage temperature:Cool
- Concentration:Total Protein: 34,7 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Purified Human Kappa Light Chain
- Purification:purified by delipidation and ion exchange chromatography
- Size:10 ml
- Pk:10 mL
Specifications
About this item
Type: Primary
Antigen: Kappa Light Chain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: