Anti-TNNI3 Mouse Monoclonal Antibody [clone: 16A12]
Catalog # BIRBORB24911-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:troponin I type 3 (cardiac)
- Antigen symbol:TNNI3
- Clonality:Monoclonal
- Clone:16A12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Western blot:Yes
- Amino acid number:85 to 92
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:8,6 mg/ml (Sigma Protein Assay Kit)
- Shipping temperature:Ice
- Immunogen:Native troponin complex.
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: TNNI3 (troponin I type 3 (cardiac))
Clonality: monoclonal
Clone: 16A12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: