Anti-INS Mouse Monoclonal Antibody (Biotin) [clone: 3A6]
BIRBORB24405-1
: Biorbyt
- Antibody type:Primary
- Antigen name:insulin
- Antigen symbol:INS
- Clonality:Monoclonal
- Clone:3A6
- Conjugation:Biotin
- Host:Mouse
- Isotype:IgG1 kappa
- Storage buffer:20 mM Tris-HCl, 150 mM Sodium Chloride, pH 7,5
- Storage temperature:Cool
- Concentration:2,1 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Purified Human Insulin
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: INS (insulin)
Clonality: monoclonal
Clone: 3A6
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: