Order Entry
Austria
ContactUsLinkComponent
Anti-PAPP-A (proMBP) Mouse Monoclonal Antibody
Anti-PAPP-A (proMBP) Mouse Monoclonal Antibody
  BIRBORB24377-1
 :  Biorbyt
Anti-PAPP-A (proMBP) Mouse Monoclonal Antibody
  BIRBORB24377-1
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen symbol:
    PAPP-A
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Western blot:
    Yes
  • Antigen synonyms:
    proMBP
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    Cool
  • Concentration:
    4,1 mg/ml (Sigma protein assay kit)
  • Shipping temperature:
    Ice
  • Immunogen:
    Purified human PAPP-A/proMBP heterotetrameric complex (htPAPP-A)
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

 

 

Type: Primary
Antigen: PAPP-A (proMBP)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: