Anti-Growth Hormone Mouse Monoclonal Antibody
BIRBORB24331-1
: Biorbyt
- Antibody type:Primary
- Antigen name:Growth Hormone
- Antigen symbol:GH
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:6,71 mg/ml (OD280)
- Shipping temperature:Ice
- Immunogen:Human Growth Hormone
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: Growth Hormone
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human