Anti-Staph. aureus Enterotoxin B Mouse Monoclonal Antibody
Catalog # BIRBORB23991-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:Staph. aureus Enterotoxin B
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:2,3 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Recombinant Staphylococcus aureus enterotoxin B expressed in E. coli.
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Staph. aureus Enterotoxin B
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: