Order Entry
Austria
ContactUsLinkComponent
Anti-Influenza A H7 Mouse Monoclonal Antibody
Anti-Influenza A H7 Mouse Monoclonal Antibody
Catalog # BIRBORB23617-1
Supplier:  Biorbyt
Anti-Influenza A H7 Mouse Monoclonal Antibody
Catalog # BIRBORB23617-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Influenza A H7
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    Cool
  • Concentration:
    2,6 mg/ml (Sigma Protein Assay)
  • Shipping temperature:
    Ice
  • Immunogen:
    Influenza A/Netherlands/219/03 (H7N7)
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: Influenza A H7
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: