Anti-Influenza A H9 Mouse Monoclonal Antibody
Catalog # BIRBORB23609-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Influenza A H9
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Recombinant hemagglutinins of H9N2 viruses: A/Chicken/Hong Kong/G9/97 (H9N2) and A/Hong Kong/1073/1999 (H9N2) strains
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Influenza A H9
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: