Anti-C. trachomatis EB Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
BIRBORB23152-1
: Biorbyt
- Antibody type:Primary
- Antigen name:C. trachomatis EB
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Storage buffer:PBS containing 10mg/ml BSA
- Storage temperature:Cool
- Concentration:1-2 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:L2 and other serovar groups
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: C. trachomatis EB
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity: