Order Entry
Austria
ContactUsLinkComponent
Anti-MHC Class Mouse Monoclonal Antibody (Biotin)
Anti-MHC Class Mouse Monoclonal Antibody (Biotin)
Supplier:  Biorbyt
Certificates
Anti-MHC Class Mouse Monoclonal Antibody (Biotin)
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB22856-500
  • BIRBORB22856-1
  • Antigen name:
  • MHC Class
  • MHC Class
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2b
  • IgG2b
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Purified mouse LN cells (C57Bl/6 anti-BALB/c).
  • Purified mouse LN cells (C57Bl/6 anti-BALB/c).
  • Purification:
  • purified by ion exchange chromatography
  • purified by ion exchange chromatography
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Frequently Bought Together