Anti-IgG Rabbit Polyclonal Antibody
Catalog # BIRBORB22313-1
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Format:antiserum
- Form:lyophilized
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Purified normal IgG isolated from pooled duck serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Secondary
Antigen: IgG (Duck) (duck IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: