Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BIRBORB21803-1
: Biorbyt
- Antibody type:Secondary
- Antigen symbol:IgM
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Goat
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) is approximately 1,6, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Purified IgM isolated from dog serum. Freunds complete adjuvant is used in the first step of the immuni- zation procedure.
- Size:1 ml
- Pk:1 mL
Type: Secondary
Antigen: IgM (Dog) (dog IgM)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: none
Reactivity: