Order Entry
Austria
ContactUsLinkComponent
Anti-Insulin Guinea pig Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-Insulin Guinea pig Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BIRBORB21788-1
Supplier:  Biorbyt
Anti-Insulin Guinea pig Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BIRBORB21788-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Insulin
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Guinea pig
  • Reactivity:
    Pig
  • Format:
    immunoconjugate
  • Form:
    lyophilized
  • UniProtKB:
    P01315
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) approximately 2,7, no foreign proteins added,
  • Shipping temperature:
    Ice
  • Tested applications:
    DOT, IEP
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

The reactivity of the antiserum is directed to insulin as tested by ELISA and immunoblotting. No reaction was obtained with other swine serum proteins.

Type: Primary
Antigen: Insulin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Guinea pig
Isotype:
Reactivity: Pig