Order Entry
Austria
ContactUsLinkComponent
Anti-lambda light chainlambda light chain Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-lambda light chainlambda light chain Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BIRBORB21755-1
Supplier:  Biorbyt
Anti-lambda light chainlambda light chain Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BIRBORB21755-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    lambda light chain
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • Format:
    immunoconjugate
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml, Fluorochrome/IgG protein ratio (F/P) approximately 2,4, no foreign proteins added,
  • Shipping temperature:
    Ice
  • Immunogen:
    A pool of purified Bence Jones lambda proteins isolated from human urine. Freunds complete adjuvant is used in the first step of the immunization procedure.
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Secondary
Antigen: lambda light chain
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: