Anti-IgG1/IgG2a/IgG2b/IgG2c Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BIRBORB21627-2
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG1/IgG2a/IgG2b/IgG2c
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Format:antiserum
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) is approximately 1,3, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Purified normal IgG isolated from pooled rat serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:2 ml
- Pk:2 mL
Specifications
About this item
Type: Secondary
Antigen: Rat IgG1 IgG2A IgG2B IgG2C
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: