Anti-L-Lactic Dehydrogenase Ldh-5(M4) Isozyme Rabbit Polyclonal Antibody
Catalog # BIRBORB21017-10
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:L-Lactic Dehydrogenase Ldh-5 (M4 Isozyme)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Storage buffer:phosphate buffered saline
- Storage temperature:Cool
- Concentration:1 mg/ml, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:L-Lactic dehydrogenase, LDH-5(M4) isoenzyme, is isolated and purified from human placenta. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:10 mg
- Pk:10 mg
Specifications
About this item
Type: Primary
Antigen: L-Lactic Dehydrogenase Ldh-5(M4) Isozyme
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human