Order Entry
Austria
ContactUsLinkComponent
Anti-L-Lactic Dehydrogenase Ldh-1(H4)/Ldh-2(H3M) Isozyme Rabbit Polyclonal Antibody
Anti-L-Lactic Dehydrogenase Ldh-1(H4)/Ldh-2(H3M) Isozyme Rabbit Polyclonal Antibody
Catalog # BIRBORB21010-10
Supplier:  Biorbyt
Anti-L-Lactic Dehydrogenase Ldh-1(H4)/Ldh-2(H3M) Isozyme Rabbit Polyclonal Antibody
Catalog # BIRBORB21010-10
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    L-Lactic Dehydrogenase Ldh-1(H4)/Ldh-2(H3M) Isozyme
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Storage buffer:
    phosphate buffered saline
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml, no foreign proteins added,
  • Shipping temperature:
    Ice
  • Immunogen:
    L-Lactic dehydrogenase, LDH-1(H4)/LDH-2(H3M) isoenzyme, is isolated and purified from human placenta. Freunds complete adjuvant is used in the first step of the immunization procedure.
  • Size:
    10 mg
  • Pk:
    10 mg

Specifications

About this item

Type: Primary
Antigen: L-Lactic Dehydrogenase Ldh-1(H4)/Ldh-2(H3M) Isozyme
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human