Anti-Oxalate Decarboxylase Rabbit Polyclonal Antibody
BIRBORB20853-10
: Biorbyt
- Antibody type:Primary
- Antigen symbol:Oxalate Decarboxylase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Storage buffer:phosphate buffered saline
- Storage temperature:Cool
- Concentration:1 mg/ml, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Oxalate decarboxylase isolated and purified from Aspergillus species. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:10 mg
- Pk:10 mg
Type: Primary
Antigen: Oxalate Decarboxylase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: